Tris(1,3-dichloro-2-propyl) Phosphate, TRC, LGC Standards
Catalog # 77695-264
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:13674-87-8
- Description:Tris(1,3-dichloro-2-propyl) Phosphate
- Size:100 mg
- Formula:C₉H₁₅Cl₆O₄P
- Molecular Weight:430.89 g/mol
- Cat. No.:77695-264