Tirofiban Hydrochloride Hydrate, TRC, LGC Standards
Catalog # 77695-226
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:150915-40-5
- Description:Tirofiban Hydrochloride Hydrate
- Size:50 mg
- Formula:C₂₂H₃₉ClN₂O₆S
- Molecular Weight:495.07 g/mol
- Cat. No.:77695-226