$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Thiamine-d3 Hydrochloride, TRC, LGC Standards
Thiamine-d3 Hydrochloride, TRC, LGC Standards
Catalog # 77695-208
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Thiamine-d3 Hydrochloride, TRC, LGC Standards
Catalog # 77695-208
Supplier:
LGC Standards
Supplier Number:
TRC-T344187-1MG
New Product
CAS Number:
Specifications
Description:
Thiamine-d3 Hydrochloride
Size:
1 mg
Cat. No.:
77695-208
Specifications
About this item
Product Details
Product Details