Pyridostigmine Bromide, TRC, LGC Standards
Catalog # 77695-057
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:101-26-8
- Description:Pyridostigmine Bromide
- Size:50 mg
- Formula:C₉H₁₃BrN₂O₂
- Molecular Weight:261.12 g/mol
- Cat. No.:77695-057