Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Psilocin O-Glucuronide, TRC, LGC Standards
Psilocin O-Glucuronide, TRC, LGC Standards
:
LGC Standards
New Product
Psilocin O-Glucuronide, TRC, LGC Standards
:
LGC Standards
New Product
CAS No.:
514212-83-0