Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Previtamin D3 (>80%), TRC, LGC Standards
Previtamin D3 (>80%), TRC, LGC Standards
77695-031
:
LGC Standards
New Product
:
Previtamin D3 (>80%), TRC, LGC Standards
77695-031
:
LGC Standards
:
TRC-P713510-1MG
New Product
:
CAS No.:
1173-13-3
Description:
Previtamin D3 (>80%)
Size:
1 mg
Cat. No.:
77695-031