Permethrin (cis/trans mixture), TRC, LGC Standards
Catalog # 77694-987
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:52645-53-1
- Description:Permethrin (cis/trans mixture)
- Size:1 g
- Formula:C₂₁H₂₀Cl₂O₃
- Molecular Weight:391.3 g/mol
- Cat. No.:77694-987