$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
N-Nitroso Hydrochlorothiazide, TRC, LGC Standards
N-Nitroso Hydrochlorothiazide, TRC, LGC Standards
Catalog # 77694-910
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
N-Nitroso Hydrochlorothiazide, TRC, LGC Standards
Catalog # 77694-910
Supplier:
LGC Standards
Supplier Number:
TRC-N542055-25MG
New Product
CAS Number:
Specifications
CAS No.:
63779-86-2
Description:
N-Nitroso Hydrochlorothiazide
Size:
25 mg
Cat. No.:
77694-910
Specifications
About this item
Product Details
Product Details