Order Entry
Puerto Rico
ContactUsLinkComponent
 
17-Methyl-androst-5-ene-3beta,17beta-diol, TRC, LGC Standards
Catalog # 77694-736
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
17-Methyl-androst-5-ene-3beta,17beta-diol, TRC, LGC Standards
Catalog # 77694-736
Supplier:  LGC Standards
Supplier Number:  TRC-M287895-5G
New Product
CAS Number:  

Specifications

  • CAS No.:
    521-10-8
  • Description:
    17-Methyl-androst-5-ene-3beta,17beta-diol
  • Size:
    5 g
  • Formula:
    C₂₀H₃₂O₂
  • Molecular Weight:
    304.47 g/mol
  • Cat. No.:
    77694-736

Specifications

About this item