Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
para-Methoxyamphetamine Hydrochloride, TRC, LGC Standards
para-Methoxyamphetamine Hydrochloride, TRC, LGC Standards
para-Methoxyamphetamine Hydrochloride, TRC, LGC Standards
77694-722
:
LGC Standards
New Product
:
para-Methoxyamphetamine Hydrochloride, TRC, LGC Standards
77694-722
:
LGC Standards
:
TRC-M260920-5MG
New Product
:
CAS No.:
50505-80-1
Description:
para-Methoxyamphetamine Hydrochloride
Size:
5 mg
Cat. No.:
77694-722