$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Meropenem-d6 (>90%), TRC, LGC Standards
Meropenem-d6 (>90%), TRC, LGC Standards
Catalog # 77694-706
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Meropenem-d6 (>90%), TRC, LGC Standards
Catalog # 77694-706
Supplier:
LGC Standards
Supplier Number:
TRC-M225617-1MG
New Product
CAS Number:
Specifications
CAS No.:
1217976-95-8
Description:
Meropenem-d6 (>90%)
Size:
1 mg
Cat. No.:
77694-706
Specifications
About this item
Product Details
Product Details