$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Mandelic Acid, TRC, LGC Standards
Mandelic Acid, TRC, LGC Standards
Catalog # 77694-685
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Mandelic Acid, TRC, LGC Standards
Catalog # 77694-685
Supplier:
LGC Standards
Supplier Number:
TRC-M162525-1G
New Product
CAS Number:
Specifications
CAS No.:
90-64-2
Description:
Mandelic Acid
Size:
1 g
Formula:
C₈H₈O₃
Molecular Weight:
152.15 g/mol
Cat. No.:
77694-685
Specifications
About this item
Product Details
Product Details