Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Loperamide-d6 Hydrochloride, TRC, LGC Standards
Loperamide-d6 Hydrochloride, TRC, LGC Standards
Loperamide-d6 Hydrochloride, TRC, LGC Standards
77694-667
:
LGC Standards
New Product
:
Loperamide-d6 Hydrochloride, TRC, LGC Standards
77694-667
:
LGC Standards
:
TRC-L469452-1MG
New Product
:
CAS No.:
1189469-46-2
Description:
Loperamide-d6 Hydrochloride
Size:
1 mg
Cat. No.:
77694-667