Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Levetiracetam-d3 (contains d0), TRC, LGC Standards
Levetiracetam-d3 (contains d0), TRC, LGC Standards
Levetiracetam-d3 (contains d0), TRC, LGC Standards
77694-656
:
LGC Standards
New Product
:
Levetiracetam-d3 (contains d0), TRC, LGC Standards
77694-656
:
LGC Standards
:
TRC-L331504-5MG
New Product
:
CAS No.:
102767-28-2
Description:
Levetiracetam-d3 (contains d0)
Size:
5 mg
Formula:
C₈H₁₄N₂O₂
Molecular Weight:
170.21 g/mol
Cat. No.:
77694-656