$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Clotiazepam (1 mg/mL in Methanol), TRC, LGC Standards
Clotiazepam (1 mg/mL in Methanol), TRC, LGC Standards
77694-639
:
LGC Standards
New Product
:
Clotiazepam (1 mg/mL in Methanol), TRC, LGC Standards
77694-639
:
LGC Standards
:
TRC-KIT2090-1X1ML
New Product
:
CAS No.:
33671-46-4
Description:
Clotiazepam (1 mg/mL in Methanol)
Size:
1x 1 ml
Cat. No.:
77694-639