Ketamine Hydrochloride (1.0 mg/mL in Methanol), TRC, LGC Standards
Catalog # 77694-629
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:1867-66-9
- Description:Ketamine Hydrochloride (1.0 mg/mL in Methanol)
- Size:5x 1 ml
- Formula:C₁₃H₁₇Cl₂NO
- Molecular Weight:274.185 g/mol
- Cat. No.:77694-629