Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Itraconazole-d5 (major), TRC, LGC Standards
Itraconazole-d5 (major), TRC, LGC Standards
77694-609
:
LGC Standards
New Product
:
Itraconazole-d5 (major), TRC, LGC Standards
77694-609
:
LGC Standards
:
TRC-I937502-10MG
New Product
:
CAS No.:
1217510-38-7
Description:
Itraconazole-d5 (major)
Size:
10 mg
Cat. No.:
77694-609