$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
rac Indapamide-D3, TRC, LGC Standards
rac Indapamide-D3, TRC, LGC Standards
Catalog # 77694-573
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
rac Indapamide-D3, TRC, LGC Standards
Catalog # 77694-573
Supplier:
LGC Standards
Supplier Number:
TRC-I500102-1MG
New Product
CAS Number:
Specifications
CAS No.:
1217052-38-4
Description:
rac Indapamide-D3
Size:
1 mg
Cat. No.:
77694-573
Specifications
About this item
Product Details
Product Details