Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Imidacloprid-d4, TRC, LGC Standards
Imidacloprid-d4, TRC, LGC Standards
Imidacloprid-d4, TRC, LGC Standards
Catalog # 77694-570
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Imidacloprid-d4, TRC, LGC Standards
Catalog # 77694-570
Supplier:
LGC Standards
Supplier Number:
TRC-I274992-2.5MG
New Product
CAS Number:
Specifications
CAS No.:
1015855-75-0
Description:
Imidacloprid-d4
Size:
2.5 mg
Cat. No.:
77694-570
Specifications
About this item
Product Details
Product Details