Order Entry
Puerto Rico
ContactUsLinkComponent
 
1,6-Hexanediol bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], TRC, LGC Standards
  77694-482
New Product
 :  
undefined
1,6-Hexanediol bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], TRC, LGC Standards
  77694-482
 :  TRC-H296885-100MG
New Product
 :  

 

  • CAS No.:
    35074-77-2
  • Description:
    1,6-Hexanediol bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]
  • Size:
    100 mg
  • Formula:
    C₄₀H₆₂O₆
  • Molecular Weight:
    638.91 g/mol
  • Cat. No.:
    77694-482