Hederacoside C, TRC, LGC Standards
Catalog # 77694-471
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:14216-03-6
- Description:Hederacoside C
- Size:50 mg
- Formula:C₅₉H₉₆O₂₆
- Molecular Weight:1221.37 g/mol
- Cat. No.:77694-471