$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Gemcitabine-13C,15N2 Hydrochloride, TRC, LGC Standards
Gemcitabine-13C,15N2 Hydrochloride, TRC, LGC Standards
:
LGC Standards
New Product
Gemcitabine-13C,15N2 Hydrochloride, TRC, LGC Standards
:
LGC Standards
New Product
CAS No.:
1262897-74-4