$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
(E)-Fluvoxamine-d4 Maleate, TRC, LGC Standards
(E)-Fluvoxamine-d4 Maleate, TRC, LGC Standards
(E)-Fluvoxamine-d4 Maleate, TRC, LGC Standards
77694-411
:
LGC Standards
New Product
:
(E)-Fluvoxamine-d4 Maleate, TRC, LGC Standards
77694-411
:
LGC Standards
:
TRC-F603503-1MG
New Product
:
CAS No.:
1432075-74-5
Description:
(E)-Fluvoxamine-d4 Maleate
Size:
1 mg
Cat. No.:
77694-411