Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Home
Standards
Organic Standards
Fipronil- 13C2 15N2, TRC, LGC Standards
Fipronil- 13C2 15N2, TRC, LGC Standards
Fipronil- 13C2 15N2, TRC, LGC Standards
Catalog # 77694-391
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Fipronil- 13C2 15N2, TRC, LGC Standards
Catalog # 77694-391
Supplier:
LGC Standards
Supplier Number:
TRC-F342202-1MG
New Product
CAS Number:
Specifications
Description:
Fipronil- 13C2 15N2
Size:
1 mg
Cat. No.:
77694-391
Specifications
About this item
Product Details
Product Details