Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Fexofenadine-D6, TRC, LGC Standards
Fexofenadine-D6, TRC, LGC Standards
Fexofenadine-D6, TRC, LGC Standards
77694-390
:
LGC Standards
New Product
:
Fexofenadine-D6, TRC, LGC Standards
77694-390
:
LGC Standards
:
TRC-F322500-1MG
New Product
:
CAS No.:
548783-71-7
Description:
Fexofenadine-D6
Size:
1 mg
Cat. No.:
77694-390