$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
N-Feruloyl Serotonin, TRC, LGC Standards
N-Feruloyl Serotonin, TRC, LGC Standards
N-Feruloyl Serotonin, TRC, LGC Standards
Catalog # 77694-388
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
N-Feruloyl Serotonin, TRC, LGC Standards
Catalog # 77694-388
Supplier:
LGC Standards
Supplier Number:
TRC-F309000-25MG
New Product
CAS Number:
Specifications
CAS No.:
68573-23-9
Description:
N-Feruloyl Serotonin
Size:
25 mg
Cat. No.:
77694-388
Specifications
About this item
Product Details
Product Details