rac Ethylphenidate Hydrochloride, TRC, LGC Standards
Catalog # 77694-361
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:19716-79-1
- Description:rac Ethylphenidate Hydrochloride
- Size:10 mg
- Formula:C₁₅H₂₂ClNO₂
- Molecular Weight:283.8 g/mol
- Cat. No.:77694-361