Dutasteride, TRC, LGC Standards
Catalog # 77694-311
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:164656-23-9
- Description:Dutasteride
- Size:100 mg
- Formula:C₂₇H₃₀F₆N₂O₂
- Molecular Weight:528.54 g/mol
- Cat. No.:77694-311