Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
3Beta-DOXYL-5alpha-cholestane, Free Radical, TRC, LGC Standards
3Beta-DOXYL-5alpha-cholestane, Free Radical, TRC, LGC Standards
3Beta-DOXYL-5alpha-cholestane, Free Radical, TRC, LGC Standards
77694-301
:
LGC Standards
New Product
:
3Beta-DOXYL-5alpha-cholestane, Free Radical, TRC, LGC Standards
77694-301
:
LGC Standards
:
TRC-D535525-5MG
New Product
:
CAS No.:
55569-61-4
Description:
3Beta-DOXYL-5alpha-cholestane, Free Radical
Size:
5 mg
Cat. No.:
77694-301