Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Diltiazem-d3 Hydrochloride, TRC, LGC Standards
Diltiazem-d3 Hydrochloride, TRC, LGC Standards
Diltiazem-d3 Hydrochloride, TRC, LGC Standards
Catalog # 77694-261
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Diltiazem-d3 Hydrochloride, TRC, LGC Standards
Catalog # 77694-261
Supplier:
LGC Standards
Supplier Number:
TRC-D460622-5MG
New Product
CAS Number:
Specifications
CAS No.:
1217623-80-7
Description:
Diltiazem-d3 Hydrochloride
Size:
5 mg
Cat. No.:
77694-261
Specifications
About this item
Product Details
Product Details