Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
10,11-Dihydro-10-hydroxy Carbamazepine-D4 (Major), TRC, LGC Standards
10,11-Dihydro-10-hydroxy Carbamazepine-D4 (Major), TRC, LGC Standards
10,11-Dihydro-10-hydroxy Carbamazepine-D4 (Major), TRC, LGC Standards
Supplier:
LGC Standards
New Product
Certificates
Expand image
10,11-Dihydro-10-hydroxy Carbamazepine-D4 (Major), TRC, LGC Standards
Supplier:
LGC Standards
New Product
About this item
Specifications
CAS No.:
1020719-39-4
Specifications