2,4-Dichlorophenoxyacetic Acid-13C6, TRC, LGC Standards
Catalog # 77694-214
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:150907-52-1
- Description:2,4-Dichlorophenoxyacetic Acid-13C6
- Size:1 mg
- Formula:Cl*C₆H₃(Cl)OCH₂COOH
- Molecular Weight:226.99 g/mol
- Cat. No.:77694-214