Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Diazinon-d10, TRC, LGC Standards
Diazinon-d10, TRC, LGC Standards
Diazinon-d10, TRC, LGC Standards
Catalog # 77694-198
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Diazinon-d10, TRC, LGC Standards
Catalog # 77694-198
Supplier:
LGC Standards
Supplier Number:
TRC-D416882-2.5MG
New Product
CAS Number:
Specifications
CAS No.:
100155-47-3
Description:
Diazinon-d10
Size:
2.5 mg
Cat. No.:
77694-198
Specifications
About this item
Product Details
Product Details