Dasatinib monohydrate, TRC, LGC Standards
Catalog # 77694-131
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:863127-77-9
- Description:Dasatinib monohydrate
- Size:1 g
- Formula:C₂₂H₂₆ClN₇O₂S·H₂O
- Molecular Weight:506.02 g/mol
- Cat. No.:77694-131