Cupriethylenediamine Hydroxide (1.0 M in H2O), TRC, LGC Standards
Catalog # 77694-106
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:14552-35-3
- Description:Cupriethylenediamine Hydroxide (1.0 M in H2O)
- Size:100 ml
- Formula:Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
- Molecular Weight:217.76 g/mol
- Cat. No.:77694-106