Cefoperazone Sodium Salt, TRC, LGC Standards
Catalog # 77694-034
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:62893-20-3
- Description:Cefoperazone Sodium Salt
- Size:5 g
- Formula:C₂₅H₂₆N₉NaO₈S₂
- Molecular Weight:667.648 g/mol
- Cat. No.:77694-034