$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
rac Carprofen, TRC, LGC Standards
rac Carprofen, TRC, LGC Standards
77694-024
:
LGC Standards
New Product
:
rac Carprofen, TRC, LGC Standards
77694-024
:
LGC Standards
:
TRC-C184350-50MG
New Product
:
CAS No.:
53716-49-7
Description:
rac Carprofen
Size:
50 mg
Formula:
C₁₅H₁₂ClNO₂
Molecular Weight:
273.72 g/mol
Cat. No.:
77694-024