Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Bupropion Hydrochloride, TRC, LGC Standards
Bupropion Hydrochloride, TRC, LGC Standards
Bupropion Hydrochloride, TRC, LGC Standards
77693-998
:
LGC Standards
New Product
:
Bupropion Hydrochloride, TRC, LGC Standards
77693-998
:
LGC Standards
:
TRC-B689625-1G
New Product
:
CAS No.:
31677-93-7
Description:
Bupropion Hydrochloride
Size:
1 g
Formula:
C₁₃H₁₉Cl₂NO
Molecular Weight:
276.20 g/mol
Cat. No.:
77693-998