N,N-Bis[3-D-gluconamidopropyl]cholamide, TRC, LGC Standards
Catalog # 77693-955
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:86303-22-2
- Description:N,N-Bis[3-D-gluconamidopropyl]cholamide
- Size:10 g
- Formula:C₄₂H₇₅N₃O₁₆
- Cat. No.:77693-955