2',4-Bis(aminophenyl)methane, TRC, LGC Standards
Catalog # 77693-951
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:1208-52-2
- Description:2',4-Bis(aminophenyl)methane
- Size:250 mg
- Formula:C₁₃H₁₄N₂
- Molecular Weight:198.264 g/mol
- Cat. No.:77693-951