$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
2-Benzothiazolethiol-d4, TRC, LGC Standards
2-Benzothiazolethiol-d4, TRC, LGC Standards
2-Benzothiazolethiol-d4, TRC, LGC Standards
Catalog # 77693-922
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
2-Benzothiazolethiol-d4, TRC, LGC Standards
Catalog # 77693-922
Supplier:
LGC Standards
Supplier Number:
TRC-B206652-5MG
New Product
CAS Number:
Specifications
CAS No.:
1346598-64-8
Description:
2-Benzothiazolethiol-d4
Size:
5 mg
Cat. No.:
77693-922
Specifications
About this item
Product Details
Product Details