Beclomethasone Dipropionate, TRC, LGC Standards
Catalog # 77693-912
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:5534-09-8
- Description:Beclomethasone Dipropionate
- Size:1 g
- Formula:C₂₈H₃₇ClO₇
- Molecular Weight:521.047 g/mol
- Cat. No.:77693-912