Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
(AlphaS,BetaR)-Bedaquiline, TRC, LGC Standards
(AlphaS,BetaR)-Bedaquiline, TRC, LGC Standards
(AlphaS,BetaR)-Bedaquiline, TRC, LGC Standards
77693-908
:
LGC Standards
New Product
:
(AlphaS,BetaR)-Bedaquiline, TRC, LGC Standards
77693-908
:
LGC Standards
:
TRC-B119540-5MG
New Product
:
CAS No.:
843663-66-1
Description:
(AlphaS,BetaR)-Bedaquiline
Size:
5 mg
Formula:
C₃₂H₃₁BrN₂O₂
Molecular Weight:
555.5 g/mol
Cat. No.:
77693-908