$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
delta7-Avenasterol (E/Z mixture), TRC, LGC Standards
delta7-Avenasterol (E/Z mixture), TRC, LGC Standards
77693-888
:
LGC Standards
New Product
:
delta7-Avenasterol (E/Z mixture), TRC, LGC Standards
77693-888
:
LGC Standards
:
TRC-A794825-0.5MG
New Product
:
CAS No.:
23290-26-8
Description:
delta7-Avenasterol (E/Z mixture)
Size:
0.5 mg
Cat. No.:
77693-888