Order Entry
Puerto Rico
ContactUsLinkComponent
(7S,9S)-9-Acetyl-6,7,9,11-tetrahydroxy-7,8,9,10-tetrahydro-5,12-naphthacenedione, TRC, LGC Standards
(7S,9S)-9-Acetyl-6,7,9,11-tetrahydroxy-7,8,9,10-tetrahydro-5,12-naphthacenedione, TRC, LGC Standards
Catalog # 77693-794
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
(7S,9S)-9-Acetyl-6,7,9,11-tetrahydroxy-7,8,9,10-tetrahydro-5,12-naphthacenedione, TRC, LGC Standards
Catalog # 77693-794
Supplier:  LGC Standards
Supplier Number:  TRC-A189248-1MG
New Product
CAS Number:  

Specifications

  • CAS No.:
    60660-75-5
  • Description:
    (7S,9S)-9-Acetyl-6,7,9,11-tetrahydroxy-7,8,9,10-tetrahydro-5,12-naphthacenedione
  • Size:
    1 mg
  • Formula:
    C₂₀H₁₆O₇
  • Molecular Weight:
    368.34 g/mol
  • Cat. No.:
    77693-794

Specifications

About this item