Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
N1-Acetylspermidine-d6 Dihydrochloride, TRC, LGC Standards
N1-Acetylspermidine-d6 Dihydrochloride, TRC, LGC Standards
N1-Acetylspermidine-d6 Dihydrochloride, TRC, LGC Standards
Catalog # 77693-789
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
N1-Acetylspermidine-d6 Dihydrochloride, TRC, LGC Standards
Catalog # 77693-789
Supplier:
LGC Standards
Supplier Number:
TRC-A187847-1MG
New Product
CAS Number:
Specifications
Description:
N1-Acetylspermidine-d6 Dihydrochloride
Size:
1 mg
Cat. No.:
77693-789
Specifications
About this item
Product Details
Product Details