Cantharidin, Mikromol, LGC Standards
Catalog # 77692-968
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:56-25-7
- Description:Cantharidin
- Size:100 mg
- Formula:C₁₀H₁₂O₄
- Molecular Weight:196.202 g/mol
- Cat. No.:77692-968