Deflazacort, Mikromol, LGC Standards
Catalog # 77692-953
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:14484-47-0
- Description:Deflazacort
- Size:250 mg
- Formula:C₂₅H₃₁NO₆
- Molecular Weight:441.52 g/mol
- Cat. No.:77692-953