Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
(R)-Citalopram Oxalate, Mikromol, LGC Standards
(R)-Citalopram Oxalate, Mikromol, LGC Standards
77692-950
:
LGC Standards
New Product
:
(R)-Citalopram Oxalate, Mikromol, LGC Standards
77692-950
:
LGC Standards
:
MM1162.03-0025
New Product
:
CAS No.:
219861-53-7
Description:
(R)-Citalopram Oxalate
Size:
25 mg
Cat. No.:
77692-950