Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
(5-Nitrofuran-2-yl)methylene Diacetate, Mikromol, LGC Standards
(5-Nitrofuran-2-yl)methylene Diacetate, Mikromol, LGC Standards
(5-Nitrofuran-2-yl)methylene Diacetate, Mikromol, LGC Standards
77692-926
:
LGC Standards
New Product
:
(5-Nitrofuran-2-yl)methylene Diacetate, Mikromol, LGC Standards
77692-926
:
LGC Standards
:
MM0943.02
New Product
:
CAS No.:
92-55-7
Description:
(5-Nitrofuran-2-yl)methylene Diacetate
Size:
100 mg
Formula:
C₉H₉NO₇
Molecular Weight:
243.17 g/mol
Cat. No.:
77692-926